Learn More
Axygen™ 48-Well PCR Microplates
Product Code. 11360175 Shop All Axygen Scientific Products
Click to view available options
Color:
Clear
Unit Size:
Pack of 10
Product Code. 11360175
Brand: Axygen™ PCR48C
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specially designed to fit most thermal cyclers
  • Volume to brim of well: 350 μL, working volume: 300 μL, volume with AxyMat: 240 μL
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance RNase, DNase, DNA-free
Format Standard
Lid Without Lid
Array 6 x 8
Color Clear
Material Polypropylene
No. of Wells 48
Sterility Non-sterile
Well Shape Round
For Use With (Application) PCR
No. per Case 50/Cs.
No. per Pack 10/Pk.