missing translation for 'onlineSavingsMsg'
Learn More

Avobenzone, MedChemExpress

Product Code. p-200101085
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16498226

Brand: Medchem Express HYB0316

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Avobenzone, a dibenzoylmethane compound, is one of the most widely used filters in sunscreens for skin photoprotection in the UVA band. Avobenzone is an endocrine disruptor that directly binds to estrogen receptor β and acts as an estrogen agonist.

Avobenzone, a dibenzoylmethane compound, is one of the most widely used filters in sunscreens for skin photoprotection in the UVA band. Avobenzone is an endocrine disruptor that directly binds to estrogen receptor β and acts as an estrogen agonist.

Specifications

Chemical Name or Material Avobenzone
CAS 70356-09-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C20H22O3
Quantity 100 mg
Solubility Information DMSO : ≥ 50 mg/mL (161.09 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(C1=CC=C(C(C)(C)C)C=C1)CC(C2=CC=C(OC)C=C2)=O
Molecular Weight (g/mol) 310.39
Formula Weight 310.39
Percent Purity 98.0%
Grade Research
Physical Form Powder
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.