missing translation for 'onlineSavingsMsg'
Learn More

Avibactam Sodium Salt, TRC

Product Code. 30009035
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009035 10 mg 10mg
30007491 5 mg 5mg
30003469 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009035 Supplier Toronto Research Chemicals Supplier No. TRCA79485010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1192491-61-4
Molecular Formula C7 H10 N3 O6 S . Na
Molecular Weight (g/mol) 287.23
Synonym Sulfuric acid, mono[(1R,2S,5R)-2-(aminocarbonyl)-7-oxo-1,6-diazabicyclo[3.2.1]oct-6-yl] ester, sodium salt (1:1), Sulfuric acid mono[(1R,2S,5R)-2-(aminocarbonyl)-7-oxo-1,6-diazabicyclo[3.2.1]oct-6-yl] ester monosodium salt, (1R,2S,5R)-7-Oxo-6-sulfooxy-1,6-diazabicyclo[3.2.1]octane-2-carboxamide sodium salt, Avibactam sodium, Avibactam sodium salt
IUPAC Name sodium;[(2S,5R)-2-carbamoyl-7-oxo-1,6-diazabicyclo[3.2.1]octan-6-yl] sulfate
SMILES [Na+].NC(=O)[C@@H]1CC[C@@H]2CN1C(=O)N2OS(=O)(=O)[O-]

Specifications

Analyte or Component Names Avibactam sodium salt
Shipping Condition Room Temperature
InChI Formula InChI=1S/C7H11N3O6S.Na/c8-6(11)5-2-1-4-3-9(5)7(12)10(4)16-17(13,14)15;/h4-5H,1-3H2,(H2,8,11)(H,13,14,15);/q;+1/p-1/t4-,5+;/m1./s1
Solution Type Neat
Industry Type Pharmaceutical
Quantity 10 mg
Formula Weight 287.0188
Percent Purity >95%
Chemical Name or Material Avibactam sodium salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.