missing translation for 'onlineSavingsMsg'
Learn More

AUZ 454, MedChemExpress

Product Code. 16463456
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
2mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16463456 25 mg 25mg
16498867 5 mg 5mg
16482407 50 mg 50mg
16470226 2 mg 2mg
16446288 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16463456 Supplier Medchem Express Supplier No. HY15004

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AUZ 454 (K03861) is a type II CDK2 inhibitor with Kd of 8.2 nM. AUZ 454 (K03861) inhibits CDK2 activity by competing with binding of activating cyclins.

AUZ 454 (K03861) is a type II CDK2 inhibitor with Kd of 8.2 nM. AUZ 454 (K03861) inhibits CDK2 activity by competing with binding of activating cyclins.

Specifications

Chemical Name or Material AUZ 454
CAS 853299-07-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C24H26F3N7O2
Quantity 25 mg
Synonym K03861
Solubility Information DMSO : 250 mg/mL (498.50 mM; Need ultrasonic)
SMILES NC1=NC(OC2=CC=C(NC(NC3=CC(C(F)(F)F)=C(CN4CCN(C)CC4)C=C3)=O)C=C2)=CC=N1
Molecular Weight (g/mol) 501.5
Formula Weight 501.5
Percent Purity 98.91%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.