missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ Clavies™ Heat-Resistant Biohazard Autoclave Gloves
Shop All SP Scienceware Products
Click to view available options
Length (English) Cuff:
5 in.
Unit Size:
Pack of 2
Description
- Ideal for removing hot objects from an autoclave or oven
- Heat-resistant up to 450°F (232°C)
- Replaces hazardous asbestos gloves
- Extended gauntlet cuff available for additional forearm protection
- Gloves are not waterproof and should not be used to handle dry ice
- Machine wash, air dry, or use the Lab-Aire™ II Glove Holder (F18931-0000) to dry on any Lab-Aire™ dryer
Specifications
Specifications
| Certifications/Compliance | CE |
| Accelerator Free | Yes |
| Applications | Laboratory, Industrial |
| Coating Material | Uncoated |
| Color | Orange |
| Latex Free | Yes |
| Material | Terrycloth |
| Product Type | Autoclave Gloves |
| Size | Universal |
| Style | Full-hand Mitt |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction