Learn More
Cole-Parmer™ Parker Hannifin™ PFA Union Connector Compression Fitting
Click to view available options
For Use With:
1/4 in. tubing O.D.
1/8 in. tubing O.D.
3/8 in. tubing O.D.
4mm tubing O.D.
6mm tubing O.D.
Shape:
Elbow
Straight
T-shape
Unit Size:
Each
Description
- Available in straight, "T", and elbow shapes for unmatched versatility
- Provided with a nut that compresses rigid tubing against the fitting body for high-pressure applications
- Excellent for use with tubing manufactured from fluoropolymers, other rigid plastics and metals
- Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal
- Sterilization through autoclaving or ethylene oxide (ETO)
Specifications
Specifications
| Autoclavable | Autoclavable |
| Material | PFA |
| For Use With | 3/8 in. tubing O.D. |
| Packaging Quantity | 1 Each |
| Sterilization Method | Autoclave or Ethylene Oxide |
| Shape | Straight |
| Product Type | Compression Fitting |