Learn More
Poulten Graf™ Autoclavable Glass Syringe
Click to view available options
Volume (Metric):
1 mL
3 mL
20 mL
30 mL
100 mL
Connection Type:
Luer Lock
Luer Tip
Unit Size:
Each
Description
- Complies with CE EN 46001 standards
- Constructed from Durobax™ glass
- With permanent amber graduation
- Acid and alkali resistant
Specifications
Specifications
| Autoclavable | Autoclavable |
| Product Type | Syringe |
| Quantity | 1/Ea. |
| Volume (Metric) | 20 mL |
| Connection Type | Luer Tip |