Learn More
Keystone Cleanroom Products Autoclavable Breather Bag for Isolators
GREENER_CHOICE
Product Code. p-7241187 Shop All Keystone Cleanroom Products Products
Change view
Click to view available options
Length (English):
6 in.
8 in.
9.5 in.
10 in.
11 in.
12 in.
13 in.
14 in.
15 in.
17 in.
18 in.
20 in.
22 in.
24 in.
26 in.
28 in.
30 in.
32 in.
34 in.
35 in.
Width (English):
2 in.
3 in.
3.5 in.
4 in.
5 in.
6 in.
7 in.
8 in.
10 in.
12 in.
14 in.
15 in.
16 in.
18 in.
24 in.
Capacity (Metric):
0.44 L
0.47 L
0.6 L
0.69 L
1.42 L
1.49 L
1.95 L
1.98 L
1.99 L
2.26 L
2.98 L
3.56 L
4.2 L
4.8 L
4.97 L
5 L
5.2 L
6.76 L
7.45 L
8.25 L
9.8 L
10.3 L
12.7 L
13.9 L
15.5 L
17.9 L
20.9 L
Unit Size:
Pack of 1000
Pack of 250
Pack of 500
31 product options available for selection
Product selection table with 31 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (English) Width (English) Capacity (Metric) unitSize
17766909 6 in. 3 in. 0.44 L Pack of 1000
17756909 9.5 in. 2 in. 0.47 L Pack of 1000
17786909 8 in. 3.5 in. 0.69 L Pack of 500
17776909 8 in. 3 in. 0.6 L Pack of 500
17716919 10 in. 5 in. 1.42 L Pack of 500
17726919 8 in. 6 in. 1.42 L Pack of 500
17796909 15 in. 4 in. 1.49 L Pack of 500
17706919 22 in. 4 in. 1.95 L Pack of 500
17736919 12 in. 6 in. 1.98 L Pack of 500
17786919 10 in. 8 in. 1.99 L Pack of 500
17776919 13 in. 7 in. 2.26 L Pack of 500
17796919 15 in. 8 in. 2.98 L Pack of 500
17726929 11 in. 10 in. 2.98 L Pack of 500
17706929 18 in. 8 in. 3.56 L Pack of 500
17746929 14 in. 12 in. 4.2 L Pack of 500
17746919 28 in. 6 in. 4.2 L Pack of 500
17756919 32 in. 6 in. 4.8 L Pack of 500
17736929 20 in. 10 in. 4.97 L Pack of 500
17756929 17 in. 12 in. 5 L Pack of 500
17766919 35 in. 6 in. 5.2 L Pack of 500
17716929 34 in. 8 in. 6.76 L Pack of 500
17786929 17 in. 16 in. 6.76 L Pack of 500
17776929 20 in. 15 in. 7.45 L Pack of 500
17766929 24 in. 14 in. 8.25 L Pack of 500
17726939 22 in. 18 in. 9.8 L Pack of 500
17796929 26 in. 16 in. 10.3 L Pack of 500
17706939 32 in. 16 in. 12.7 L Pack of 250
17716939 35 in. 16 in. 13.9 L Pack of 250
17736939 26 in. 24 in. 15.5 L Pack of 250
17746939 30 in. 24 in. 17.9 L Pack of 250
17756939 35 in. 24 in. 20.9 L Pack of 250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 31 options available.
31 options
Product Code. 17766909 Supplier Keystone Cleanroom Products Supplier No. BBMGTVKSS0306HH
Only null left
Add to basket
Add to basket
These self-sealing breather bags offer a chevron peel and hang hole to suspend previously steam sterilized pouches in an isolator for VHP decontamination of outer packaging.
  • Tyvek™ and polyethylene film construction for high strength and exceptional microbial barrier does not absorb or adsorb VHP
  • Designed for steam sterilization, but also works with gamma irradiation, EtO and VHP
  • Fiber-free peelable seals
  • High clarity for product visibility
  • Recommended for sterilization of pharmaceutical equipment
  • Manufactured and packaged in a certified ISO 6 cleanroom
  • Works with both Impulse and Continuous Band Sealers
  • Improves the efficiency of Parts Preparation Department
Specifications
Autoclavable Autoclavable
Certifications/Compliance COC
Closure Type Self-seal
Color White
Disposable Single-use
For Use With (Equipment) Isolator
Material Tyvek
Sterility Non-sterile
Latex Free Yes
Sterilization Method Steam, EtO, Gamma Irradiation, VHP
Thickness (English) 8 mil
Length (English) 6 in.
Width (English) 3 in.
Product Type Autoclave Bag
Capacity (English) 15.1 oz.
Capacity (Metric) 0.44 L
Length (Metric) 15.2 cm
Width (Metric) 7.6 cm
Thickness (Metric) 203 μm