Learn More
3M™ Aura™ Particulate Respirators 9300+ Series
Product Code. 17217890
Click to view available options
Filter Type:
FFP1
FFP2
FFP3
With Exhalation Valve:
No
Yes
Packaging Type:
Bulk
Individually Packaged
Unit Size:
Pack of 10
Pack of 20
Pack of 440
Product Code. 17217890
Brand: 3M™ 7100241313
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Particulate respirators 9300+ Series feature an ergonomic, flat-fold, 3-panel design with a low breathing resistance filter media. Available in FFP1, FFP2 and FFP3 protection levels with valved and unvalved models, respirators have an innovative chin tab for easy adjustments and an embossed, sculpted upper panel to help reduce eyewear fogging.
  • Provide comfortable protection against dusts and mists in a wide variety of industrial applications and other hazardous environments.
  • Offers FFP1, FFP2 and FFP3 protection.
  • 3-panel design allows for greater facial movement.
  • Sculpted, low-profile style of the embossed upper panel creates an effective seal, reduces eyewear fogging and improves eyewear compatibility.
  • Low resistance filter technology promotes easier breathing and the innovative chin tab allows simple adjustments.
  • Choose without valve style or with valve styles that feature 3M™ Cool Flow™ to reduce heat build-up and help provide comfortable protection in hot and humid conditions.
  • Individually packed in hygienic wrapping to prevent contamination before use.
  • Braided, colour-coded headbands help you identify the protection level offered: yellow for FFP1 (APF 4), blue for FFP2 (APF 10) and red for FFP3 (APF 20).
  • Foldable design makes them easy to store when not in use.
  • Respirators work well with 3M eyewear and hearing protection products.
TRUSTED_SUSTAINABILITY
Specifications
Assigned Protection Factor (APF) 10
Certifications/Compliance (Europe) European Standard EN 149 for Filtering Half Masks to Protect Against Particles
Certifications (EN) EN 149
Description Unvalved, Bulk
Color White
Design Type Flat-fold
Disposable Single-use
Filter Type FFP2
Material Polypropylene
Product Type General Purpose Particulate Respirator
Size One Size
With Exhalation Valve No
Filtration Layer Material Polypropylene
Latex Free Yes
Material Headstrap Polyisoprene
Model 9320D+
Nose Piece Material Aluminum, Polyurethene
Nosepiece Type Sculpted
Packaging Type Bulk
Quantity 440/Cs.
Resistant To Dusts and mists found in a wide variety of industrial applications and other work situations
Shape Cup
Sterility Non-sterile
Style 3-panel
Strap Attachment Type Colour-coded Headband, Blue
FDA Cleared Surgical Mask No