Learn More
Aucubine, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Aucubin |
| CAS | 479-98-1 |
| Molecular Formula | C15 H22 O9 |
| Quantity | 10 mg |
| Synonym | β-D-Glucopyranoside, (1S,4aR,5S,7aS)-1,4a,5,7a-tetrahydro-5-hydroxy-7-(hydroxymethyl)cyclopenta[c]pyran-1-yl, Aucubin (6CI,7CI,8CI), β-D-Glucopyranoside, 1,4a,5,7a-tetrahydro-5-hydroxy-7-(hydroxymethyl)cyclopenta[c]pyran-1-yl, [1S-(1α,4aα,5α,7aα)]-, (1S,4aR,5S,7aS)-1,4a,5,7a-Tetrahydro-5-hydroxy-7-(hydroxymethyl)cyclopenta[c]pyran-1-yl β-D-glucopyranoside, Acubin, Aucubine, Aucubosid, Aucuboside, Rhimantin |
| InChI Formula | InChI=1S/C15H22O9/c16-4-6-3-8(18)7-1-2-22-14(10(6)7)24-15-13(21)12(20)11(19)9(5-17)23-15/h1-3,7-21H,4-5H2/t7-,8+,9+,10+,11+,12-,13+,14-,15-/m0/s1 |
| SMILES | OC[C@H]1O[C@@H](O[C@@H]2OC=C[C@H]3[C@H](O)C=C(CO)[C@@H]23)[C@H](O)[C@@H](O)[C@@H]1O |
| IUPAC Name | (2S,3R,4S,5S,6R)-2-[[(1S,4aR,5S,7aS)-5-hydroxy-7-(hydroxymethyl)-1,4a,5,7a-tetrahydrocyclopenta[c]pyran-1-yl]oxy]-6-(hydroxymethyl)oxane-3,4,5-triol |
| Molecular Weight (g/mol) | 346.33 |
| Formula Weight | 346.1264 |