Learn More
Aucubin, MedChemExpress
Product Code. 16464867
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. 16464867
Brand: Medchem Express HYN0664
MedChemExpress Aucubin, an iridoid glucoside, is isolated from Plantago asiatica, Eucommia ulmoides, the leaves of Aucuba japonica and more recently from butterfly larva. Aucubin has many biological activities, such as antioxidant, anti-aging, anti-inflammatory, antimicrobial, anti-fibrotic, anti-cancer, hepatoprotective, neuroprotective and osteoprotective effects.
Aucubin, an iridoid glucoside, is isolated from Plantago asiatica, Eucommia ulmoides, the leaves of Aucuba japonica and more recently from butterfly larva. Aucubin has many biological activities, such as antioxidant, anti-aging, anti-inflammatory, antimicrobial, anti-fibrotic, anti-cancer, hepatoprotective, neuroprotective and osteoprotective effects.
Specifications
Chemical Name or Material Aucubin
CAS 479-98-1
Color White
Purity Grade Notes Research
Molecular Formula C15H22O9
Quantity 50 mg
Solubility Information DMSO : 100 mg/mL (288.74 mM; Need ultrasonic)
SMILES O[C@H]([C@@H](O)[C@@H]1O)[C@](O[C@@H]1CO)([H])O[C@H](OC=C2)[C@@]3([H])[C@]2([H])[C@H](O)C=C3CO
Molecular Weight (g/mol) 346.33
Formula Weight 346.33
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus