missing translation for 'onlineSavingsMsg'
Learn More

Atuveciclib, MedChemExpress

Product Code. p-200099999
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16468137 5 mg 5mg
16425428 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16468137 Supplier Medchem Express Supplier No. HY12871B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Atuveciclib (BAY-1143572) is a potent and highly selective, oral PTEFb/CDK9 inhibitor. Atuveciclib (BAY-1143572) inhibits CDK9/CycT1 with an IC50 of 13 nM.

Atuveciclib (BAY-1143572) is a potent and highly selective, oral PTEFb/CDK9 inhibitor. Atuveciclib (BAY-1143572) inhibits CDK9/CycT1 with an IC50 of 13 nM.

Specifications

Chemical Name or Material Atuveciclib
Color White
Purity Grade Notes Research
Molecular Formula C18H18FN5O2S
Quantity 5 mg
Synonym BAY-1143572
Solubility Information DMSO : ≥ 128.5 mg/mL (331.67 mM)
SMILES N=[S@](CC1=CC(NC2=NC(C3=CC=C(F)C=C3OC)=NC=N2)=CC=C1)(C)=O
Molecular Weight (g/mol) 387.43
Formula Weight 387.43
Percent Purity 99.2%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.