Learn More
Invitrogen™ Attune™ NxT External Fluid Supply
Click to view available options
Unit Size:
Each
Description
The Attune™ NxT External Fluid Supply is an accessory for the Attune™ NxT Acoustic Focusing Cytometer. It was designed to facilitate the use of a larger buffer and waste container. The system works by monitoring and supplying the internal fluid tanks on the Attune NxT instrument so that the system can run without having to stop instrument operation to change out the fluid tanks or empty the waste. The external fluid system uses the 10 L cube of Attune™ Focusing Fluid (Cat. No. A24904) and a Nalgene™ waste bottle. The external fluid supply is ideal for use with the Attune™ NxT Auto Sampler and enables the collection of approximately 40 96-well or 10 384-well plates without changing the fluid tanks.
Specifications
Specifications
| For Use With (Equipment) | Attune™ NxT Acoustic Focusing Cytometer |
| Product Line | Attune |
| Shipping Condition | Room Temperature |
| Product Type | Flow Cytometer Accessory |