missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Atrial Natriuretic Peptide (1-29), Chicken
Click to view available options
Quantity:
0.5 mg
1 mg
Unit Size:
0.5mg
1mg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Species | Chicken |
| Purification Method | HPLC |
| Content And Storage | -30°C to -10°C |
| Format | Lyophilized Powder |
| Molecular Weight (g/mol) | 3160.61 |
| Product Type | Atrial Natriuretic Peptide (1-29), chicken |
| Quantity | 1 mg |
| Regulatory Status | RUO - research use only |
For Research Use Only
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido