missing translation for 'onlineSavingsMsg'
Learn More

Atrasentan hydrochloride, MedChemExpress

Product Code. 16456488
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16456488 10 mg 10mg
16442587 50 mg 50mg
16419077 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16456488 Supplier Medchem Express Supplier No. HY15403A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Atrasentan hydrochloride (ABT-627 hydrochloride) is a selective endothelin A receptor antagonist with an IC50 of 0.0551 nM for ETA.

Atrasentan hydrochloride (ABT-627 hydrochloride) is a selective endothelin A receptor antagonist with an IC50 of 0.0551 nM for ETA.

Specifications

Chemical Name or Material Atrasentan hydrochloride
CAS 195733-43-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C29H39ClN2O6
Quantity 10 mg
Synonym ABT-627 hydrochloride(+)-A 127722 hydrochlorideA-147627 hydrochloride
Solubility Information DMSO : ≥ 100 mg/mL (182.79 mM) ∣H2O : 0.5 mg/mL (0.91 mM; ultrasonic and warming and adjust pH to 4 with HCl and heat to 60°C) ∣0.1 M HCL : < 1 mg/mL (ultrasonic;warming;adjust pH to 1 with HCl;heat to 60°C) (insoluble)
SMILES O=C([C@H]1[C@H](C2=CC=C(OC)C=C2)N(CC(N(CCCC)CCCC)=O)C[C@@H]1C3=CC=C(OCO4)C4=C3)O.Cl
Molecular Weight (g/mol) 547.08
Formula Weight 547.08
Percent Purity 99.51%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.