Learn More
Atractyloside Dipotassium, TRC
Specifications
Specifications
| Chemical Name or Material | Atractyloside Dipotassium |
| CAS | 102130-43-8 |
| Molecular Formula | C30 H46 O16 S2 . 2K |
| Quantity | 1 mg |
| Synonym | (2beta,15alpha)-15-Hydroxy-2-[[2-O-(3-methyl-1-oxobutyl)-3,4-di-O-sulfo-beta-D-glucopyranosyl]oxy]-19-norkaur-16-en-18-oic Acid Dipotassium |
| InChI Formula | InChI=1S/C30H46O16S2.2K/c1-14(2)9-22(32)44-25-24(46-48(39,40)41)23(45-47(36,37)38)20(13-31)43-28(25)42-17-10-18(27(34)35)19-7-8-30-11-16(15(3)26(30)33)5-6-21(30)29(19,4)12-17;;/h14,16-21,23-26,28,31,33H,3,5-13H2,1-2,4H3,(H,34,35)(H,36,37,38)(H,39,40,41);;/q;2*+1/p-2/t16-,17-,18-,19-,20?,21+,23-,24?,25?,26+,28-,29-,30-;;/m1../s1 |
| SMILES | CC(C)CC(=O)OC1[C@H](O[C@@H]2C[C@H]([C@H]3CC[C@@]45C[C@@H](CC[C@H]4[C@]3(C)C2)C(=C)[C@@H]5O)C(=O)O)OC(CO)[C@@H](OS(=O)(=O)O[K])C1OS(=O)(=O)O[K] |
| IUPAC Name | [(2R,5R)-2-[[(1R,4R,5R,7R,9R,10S,13R,15S)-5-carboxy-15-hydroxy-9-methyl-14-methylidene-7-tetracyclo[11.2.1.01,10.04,9]hexadecanyl]oxy]-6-(hydroxymethyl)-3-(3-methylbutanoyloxy)-5-potassiooxysulfonyloxyoxan-4-yl]oxysulfonyloxypotassium |
| Molecular Weight (g/mol) | 802.987 |
| Formula Weight | 802.134 |