Learn More
Atractylodin, MedChemExpress
Product Code. 16498908
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16498908 10 mg 10mg
16421358 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16498908 Supplier Medchem Express Supplier No. HYN0238
MedChemExpress Atractylodin (Atractydin) is an active component of the essential oil contained in the rhizomes of Atractylodes lancea and A. chinensis. Atractylodin is natural insecticide and is active against Tribolium castaneum.
Atractylodin (Atractydin) is an active component of the essential oil contained in the rhizomes of Atractylodes lancea and A. chinensis. Atractylodin is natural insecticide and is active against Tribolium castaneum.
Specifications
Chemical Name or Material Atractylodin
Physical Form Solid
CAS 55290-63-6
Color Light Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C13H10O
Synonym Atractydin
Solubility Information DMSO : ≥ 6 mg/mL (32.93 mM)
SMILES C/C=C/C#CC#C/C=C/C1=CC=CO1
Molecular Weight (g/mol) 182.22
Formula Weight 182.22
Percent Purity 98.09%
Grade Research