missing translation for 'onlineSavingsMsg'
Learn More

Atorvastatin Cyclic (Fluorophenyl) Sodium Salt Impurity, TRC

Product Code. 17965095
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17965095 1 mg 1mg
17924285 5 mg 5mg
17923825 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17965095 Supplier Toronto Research Chemicals Supplier No. TRCA7918201MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1315629-79-8
Molecular Formula C33 H34 F N2 O7 . Na
Molecular Weight (g/mol) 612.62
Synonym 1b-(4-Fluorophenyl)hexahydro-β,7-dihydroxy-7-(1-methylethyl)-1a-phenyl-7a-[(phenylamino)carbonyl]-3H-oxireno[3,4]pyrrolo[2,1-b][1,3]oxazine-3-butanoic Acid Sodium Salt (1:1)
IUPAC Name sodium;4-[1b-(4-fluorophenyl)-7-hydroxy-1a-phenyl-7a-(phenylcarbamoyl)-7-propan-2-yl-4,5-dihydro-3H-oxireno[1,2]pyrrolo[3,5-b][1,3]oxazin-3-yl]-3-hydroxybutanoate
SMILES [Na+].CC(C)C1(O)N2CCC(CC(O)CC(=O)[O-])OC2(c3ccc(F)cc3)C4(OC14C(=O)Nc5ccccc5)c6ccccc6

Specifications

Analyte or Component Names Atorvastatin Cyclic (Fluorophenyl) Sodium Salt Impurity
Shipping Condition Room Temperature
InChI Formula InChI=1S/C33H35FN2O7.Na/c1-21(2)32(41)31(29(40)35-25-11-7-4-8-12-25)30(43-31,22-9-5-3-6-10-22)33(23-13-15-24(34)16-14-23)36(32)18-17-27(42-33)19-26(37)20-28(38)39;/h3-16,21,26-27,37,41H,17-20H2,1-2H3,(H,35,40)(H,38,39);/q;+1/p-1
Solution Type Neat
Industry Type Forensics
Quantity 1 mg
Formula Weight 612.2248
Chemical Name or Material Atorvastatin Cyclic (Fluorophenyl) Sodium Salt Impurity
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.