missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Atlantic Blue™ Anti-Human CD18 (TS1/18), Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech AB-65190-100TESTS
This item is not returnable.
View return policy
Description
The CD18 antibody from Proteintech is a mouse monoclonal antibody generated with human beta 2 integrin of human CD18. This antibody recognizes human antigen. The CD18 antibody has been validated for the following applications: FC analysis. This antibody is atlantic blue™.Specifications
| CD18 | |
| Monoclonal | |
| 5 μl/test | |
| CD18; LAD; LCAMB; LFA-1; MAC-1; MF17; MFI7 | |
| Human Beta 2 Integrin | |
| 100 Tests | |
| Primary | |
| Human | |
| Flow Antibody | |
| IgG1 κ |
| Flow Cytometry | |
| TS1/18 | |
| Atlantic Blue | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 3689 | |
| Store at 2-8°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction