Learn More
Vacuubrand ATEX Vacuum gauge head DMP 331i, BDSENSORS, with KF DN 16, range: 1000-1 mbar, accuracy +-1 mbar
Brand: Vacuubrand 20635423
ATEX Vacuum gauge head DMP 331i, BDSENSORS, with KF DN 16, range: 1000-1 mbar, accuracy +-1 mbar