missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATAD1 Monoclonal Antibody (S125-10), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA527603
This item is not returnable.
View return policy
Description
ATAD1 Monoclonal antibody specifically detects ATAD1 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.
Specifications
| ATAD1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.09% sodium azide; pH 7.4 | |
| Q505J9, Q8NBU5, Q9D5T0 | |
| ATAD1 | |
| Fusion protein amino acids 1-361 (Full length) of mouse Thorase | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| S125-10 | |
| Unconjugated | |
| ATAD1 | |
| 4921525H23Rik; AAA+ ATPase Thorase; Ab2-088; AFDC1; Atad1; ATPase family AAA domain containing 1; ATPase family AAA domain-containing protein 1; ATPase family, AAA domain containing 1; ATPase Thorase; AW107648; FLJ14600; FNP001; neuroprotective protein 6; Npg6; Thorase | |
| Mouse | |
| Protein G | |
| RUO | |
| 309532, 67979, 84896 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction