missing translation for 'onlineSavingsMsg'
Learn More

AT13148, MedChemExpress

Product Code. 16456978
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16456978

Brand: Medchem Express HY16071

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AT13148 is an orally active and ATP-competitive, multi-AGC kinase inhibitor with IC50s of 38 nM/402 nM/50 nM, 8 nM, 3 nM, and 6 nM/4 nM for Akt1/2/3, p70S6K, PKA, and ROCKI/II, respectively.

AT13148 is an orally active and ATP-competitive, multi-AGC kinase inhibitor with IC50s of 38 nM/402 nM/50 nM, 8 nM, 3 nM, and 6 nM/4 nM for Akt1/2/3, p70S6K, PKA, and ROCKI/II, respectively.

Specifications

Chemical Name or Material AT13148
Physical Form Powder
CAS 1056901-62-2
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C17H16ClN3O
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 50 mg/mL (159.35 mM; Need ultrasonic)
SMILES ClC1=CC=C([C@](C2=CC=C(C3=CNN=C3)C=C2)(O)CN)C=C1
Molecular Weight (g/mol) 313.78
Formula Weight 313.78
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.