Learn More
Saint-Gobain AstiPure™ RC1 Series PFA Union Panel Mount
Product Code. 11964139
Click to view available options
:
Length: 96mm
Product Code. 11964139
Brand: Saint-Gobain RC11013
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Material PFA
Overall Length (Metric) 96 mm
For Use With For 10 x 13mm dia. tubing
Packaging Quantity 1 Each
Port Size 21.5 x 10mm
Product Type Union Panel Mount