Learn More
Saint-Gobain AstiPure™ CAR Series PFA Check Valve
Click to view available options
Diameter (Metric) Inner:
5 mm
Unit Size:
Each
Specifications
Specifications
| Diameter (Metric) Inner | 5 mm |
| Material | PFA |
| Overall Length (Metric) | 91 mm |
| For Use With | For 6 x 8mm dia. tubing |
| Packaging Quantity | 1 Each |
| Maximum Pressure | 0.05 bar |
| Product Type | Check Valve |