Learn More
Honeywell™ Ascender ESD Safety Footwear
Product Code. 15339643
Change view
Click to view available options
Size:
44
Unit Size:
1 pair
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15339643 44 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15339643 Supplier Honeywell™ Supplier No. 62 438 57 44
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Certifications/Compliance EN ISO 20345: 2011 S3 CI SRC, EN 61340-4-3:2001
Liner Material 3D
Material Leather, Textile
Size 44
Resistant To Water
Waterproof Yes
Product Type Safety Shoe