Learn More
ASB6 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21449-1-AP-20UL
This item is not returnable.
View return policy
Specifications
| ASB6 | |
| Polyclonal | |
| Unconjugated | |
| ASB6 | |
| ASB 6, ASB6 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 140459 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.34 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9NWX5 | |
| ASB6 | |
| ASB6 Fusion Protein Ag13863 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |