missing translation for 'onlineSavingsMsg'
Learn More

Artesunate, MedChemExpress

Product Code. p-200098177
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16479026 100 mg 100mg
16434787 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16479026 Supplier Medchem Express Supplier No. HYN0193

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Artesunate is an inhibitor of both STAT-3 and exported protein 1 (EXP1).

Artesunate is an inhibitor of both STAT-3 and exported protein 1 (EXP1).

Specifications

Chemical Name or Material Artesunate
CAS 88495-63-0
Color White
Purity Grade Notes Research
Molecular Formula C19H28O8
Quantity 100 mg
Solubility Information DMSO : 83.33 mg/mL (216.77 mM; Need ultrasonic) ∣7.5% sodium bicarbonate : 20 mg/mL (52.03 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(O[C@H]1[C@H](C)[C@]2([H])CC[C@@H](C)[C@]3([H])CC[C@@](O4)(C)OO[C@]32[C@]4([H])O1)CCC(O)=O
Molecular Weight (g/mol) 384.42
Formula Weight 384.42
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.