missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARPP-19 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11678-1-AP-20UL
This item is not returnable.
View return policy
Description
Protein phosphatase inhibitor that specifically inhibits protein phosphatase 2A (PP2A) during mitosis. When phosphorylated at Ser-62 during mitosis, specifically interacts with PPP2R2D (PR55-delta) and inhibits its activity, leading to inactivation of PP2A, an essential condition to keep cyclin-B1-CDK1 activity high during M phase. May indirectly enhance GAP-43 expression by binding to the NGF-regulatory region of its mRNA.Specifications
| ARPP-19 | |
| Polyclonal | |
| Unconjugated | |
| ARPP19 | |
| ARPP 16, ARPP 19, ARPP16, ARPP19, ARPP-19 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10776, 59046, 60336 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.51 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P56211, P56212, Q712U5 | |
| Arpp19 | |
| ARPP-19 Fusion Protein Ag2279 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction