Learn More
Invitrogen™ ARPC5 Recombinant Rabbit Monoclonal Antibody (SR34-02)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA532019
Description
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
This gene encodes one of seven subunits of the human Arp2/3 protein complex. The Arp2/3 protein complex has been implicated in the control of actin polymerization in cells and has been conserved through evolution. The exact role of the protein encoded by this gene, the p16 subunit, has yet to be determined.
Specifications
| ARPC5 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4 | |
| O15511, Q4KLF8, Q9CPW4 | |
| ARPC5 | |
| Synthetic peptide within Human p16 ARC aa 1-50. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot | |
| SR34-02 | |
| Unconjugated | |
| ARPC5 | |
| 5830443F10Rik; actin related protein 2/3 complex subunit 5; actin related protein 2/3 complex subunit 5, 16kDa; actin related protein 2/3 complex, subunit 5; actin related protein 2/3 complex, subunit 5, 16kDa; actin-related protein 2/3 complex subunit 5; actin-related protein 2/3 complex subunit 5-like protein; ARC16; Arp2/3 complex 16 kDa subunit; Arp2/3 protein complex subunit p16; Arpc5; dJ127C7.3; p16-ARC | |
| Rabbit | |
| Protein A | |
| RUO | |
| 10092, 360854, 67771 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.