missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ARL6IP6 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15741171 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15741171 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15741171 Supplier Invitrogen™ Supplier No. PA553227

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: MSFAESGWRS ALRRRGPGTP GPVARPSYSS FTQGDSWGEG EVDEEEGCDQ VARDLRAEFS AGAWSEPRKR SVLPPDGNGS PVLPDKRNGI FP Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 67%.

The ARL6IP6 gene has been identified as a novel candidate gene associated with a syndromic form of Cutis Marmorata Telangiectatica Congenita (CMTC), suggesting that ischemic stroke or transient ischemic attacks (TIA) might represent the mild end of the phenotypic spectrum, with autosomal recessive CMTC as the severe manifestation. The ARL6IP6 influences the susceptibility to these conditions, highlighting its importance in vascular and dermatological syndromes. Mutations in ARL6IP6 can lead to a range of phenotypic outcomes, from mild ischemic events to serious congenital skin disorders, emphasizing the need for deeper genetic and phenotypic exploration to understand its role in human pathology. Moreover, the gene is potentially involved in intracellular trafficking processes that are crucial for proper cellular function, which can be impacted by genetic variations, leading to clinical manifestations.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARL6IP6
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARL6IP6
Gene Accession No. Q8N6S5
Gene Alias 2310057C01Rik; 2610529A11Rik; ADP ribosylation factor like GTPase 6 interacting protein 6; ADP-ribosylation factor GTPase 6 interacting protein 6; ADP-ribosylation factor like GTPase 6 interacting protein 6; ADP-ribosylation factor-like 6 interacting protein 6; ADP-ribosylation factor-like protein 6-interacting protein 6; ADP-ribosylation-like factor 6 interacting protein 6; ADP-ribosylation-like factor 6-interacting protein 6; AIP6; aip-6; ARL-6-interacting protein 6; Arl6ip6; PFAAP1; Phosphonoformate immuno-associated protein 1; regulated by phosphonoformate
Gene Symbols ARL6IP6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ARL6IP6. Recombinant protein control fragment (Product #RP-91120).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 151188
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.