Learn More
Arimistane, TRC
Product Code. 30135923
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135923 10 g 10g
30132508 5 g 5g
30008474 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30135923 Supplier Toronto Research Chemicals Supplier No. TRCA79489810G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1420-49-1
Molecular Formula C19H24O2
Molecular Weight (g/mol) 284.39
Synonym Androsta-3,5-diene-7,17-dione, Arimistane, NSC 134910, Androst-3,5-diene-7,17-dione
IUPAC Name (8R,9S,10R,14S)-10,13-dimethyl-2,8,9,11,12,14,15,16-octahydro-1H-cyclopenta[a]phenanthrene-7,17-dione
SMILES CC12CC[C@H]3[C@H]([C@@H]1CCC2=O)C(=O)C=C4C=CCC[C@]34C
Specifications
Quantity 10 g
InChI Formula InChI=1S/C19H24O2/c1-18-9-4-3-5-12(18)11-15(20)17-13-6-7-16(21)19(13,2)10-8-14(17)18/h3,5,11,13-14,17H,4,6-10H2,1-2H3/t13-,14-,17-,18-,19?/m0/s1
Formula Weight 284.1776
Percent Purity >95%
Chemical Name or Material Androst-3,5-diene-7,17-dione
Grade HPLC