missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARHGDIA Monoclonal Antibody (OTI1F2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501089
This item is not returnable.
View return policy
Description
Aplysia Ras-related homologs (ARHs), also called Rho genes, belong to the RAS gene superfamily encoding small guanine nucleotide exchange (GTP/GDP) factors. The ARH proteins may be kept in the inactive, GDP-bound state by interaction with GDP dissociation inhibitors, such as ARHGDIA.Specifications
| ARHGDIA | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P52565, Q5XI73 | |
| ARHGDIA | |
| Full length human recombit protein of human ARHGDIA produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Canine, Human, Rat | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Western Blot | |
| OTI1F2 | |
| Unconjugated | |
| ARHGDIA | |
| GDIA1, HEL-S-47e, NPHS8, RHOGDI, RHOGDI-1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 360678, 396, 475924 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction