missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARHGAP39 Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81940MFV500
This item is not returnable.
View return policy
Description
ARHGAP39 Polyclonal antibody specifically detects ARHGAP39 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications
| ARHGAP39 | |
| Polyclonal | |
| Western Blot, ELISA | |
| CrGAP, CrossGAP Homolog, CrossGAP Homolog (Drosophila), KIAA1688, Rho GTPase activating protein 39, RhoGAP93B Homolog, RhoGAP93B Homolog (Drosophila), Vilse | |
| ARHGAP39 antibody was raised against an 18 amino acid synthetic peptide near the carboxy terminus of human ARHGAP39. The immunogen is located within amino acids 880 - 930 of ARHGAP39. Amino Acid Squence: MGMQRERYPERQLPWVQT | |
| 0.1 mL | |
| Neuroscience | |
| 80728 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction