Learn More
Thermo Scientific™ [Arg0] Met-Enkephalin
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Species | Rat |
| Purification Method | HPLC |
| Content And Storage | -30°C to -10°C |
| Format | Solid |
| Molecular Weight (g/mol) | 729.84 |
| Product Type | [Arg0] Met-Enkephalin |
| Quantity | 1 mg |
| Regulatory Status | RUO - research use only |
| Sequence | Arg-Tyr-Gly-Gly-Phe-Met |
For Research Use Only