missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARD1B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15670-1-AP-20UL
This item is not returnable.
View return policy
Description
Displays alpha (N-terminal) acetyltransferase activity. Proposed alternative catalytic subunit of the N-terminal acetyltransferase A (NatA) complex.Specifications
| ARD1B | |
| Polyclonal | |
| Unconjugated | |
| NAA11 | |
| ARD1 homolog B (S. cerevisiae), ARD1B, ARD2, hARD2, NAA11 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 289482, 84779, 97243 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q3UX61, Q4V8K3, Q9BSU3 | |
| Naa11 | |
| ARD1B Fusion Protein Ag8135 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction