Learn More
Thermo Scientific™ Arctic Express™ Dual Storage Systems
Product Code. p-4530741 Shop All Thermo Scientific Products
Click to view available options
Capacity (Metric):
10 L
28 L
Unit Size:
Each
Product Code. 10338253
Brand: Thermo Scientific™ CK50920
Store and ship samples with Arctic Express dual storage systems, which feature hard-shell shipping containers.
Protect nonhazardous samples during shipment and storage using Thermo Scientific™ Arctic Express™ Storage Systems. Featuring excellent flexibility and safety features, these systems can be used as dry shippers or laboratory cryo vessels.
Features:
  • When filled with liquid nitrogen and placed in the laboratory, unit can be used as a “can and cane” storage system indefinitely with static holding times of up to 125 days.
  • Aluminum vessels come with lockable lid and hold six numbered, color-coded, stainless-steel cans (included), which accommodate canes that hold ampules.
TRUSTED_SUSTAINABILITY
Specifications
Type Manual Fill LN2 Dewar
Capacity (Metric) 10 L
No. of Doors 1
Model Arctic Express Dual 10
Static Holding Time 21 shipper/42 lab
Diameter (English) 10.2 in.
Diameter (English) Inside Neck 2 in.
Diameter (Metric) 25.9 cm
Diameter (Metric) Inside Neck 5.08 cm
Vial Capacity 150 x 2mL