missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Archaeosine Acetate Salt, TRC
Specifications
Specifications
| Chemical Name or Material | Archaeosine Acetate Salt |
| Analyte or Component Names | Archaeosine Acetate Salt |
| Shipping Condition | Blue Ice |
| Molecular Formula | C12 H16 N6 O5 . C2 H4 O2 |
| InChI Formula | InChI=1S/C12H16N6O5.C2H4O2/c13-8(14)3-1-18(9-5(3)10(22)17-12(15)16-9)11-7(21)6(20)4(2-19)23-11;1-2(3)4/h1,4,6-7,11,19-21H,2H2,(H3,13,14)(H3,15,16,17,22);1H3,(H,3,4)/t4-,6-,7-,11-;/m1./s1 |
| Solution Type | Neat |
| SMILES | CC(=O)O.NC(=N)c1cn([C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O)c3N=C(N)NC(=O)c13 |
| IUPAC Name | acetic acid;2-amino-7-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-4-oxo-3H-pyrrolo[2,3-d]pyrimidine-5-carboximidamide |
| Industry Type | Pharmaceutical |
| Molecular Weight (g/mol) | 384.345 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?