missing translation for 'onlineSavingsMsg'
Learn More

Archaeosine Acetate Salt, TRC

Product Code. 30344831
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30344831 1 mg 1mg
30347159 5 mg 5mg
30332847 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30344831 Supplier Toronto Research Chemicals Supplier No. TRCA7666601MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Archaeosine Acetate Salt
Analyte or Component Names Archaeosine Acetate Salt
Shipping Condition Blue Ice
Molecular Formula C12 H16 N6 O5 . C2 H4 O2
InChI Formula InChI=1S/C12H16N6O5.C2H4O2/c13-8(14)3-1-18(9-5(3)10(22)17-12(15)16-9)11-7(21)6(20)4(2-19)23-11;1-2(3)4/h1,4,6-7,11,19-21H,2H2,(H3,13,14)(H3,15,16,17,22);1H3,(H,3,4)/t4-,6-,7-,11-;/m1./s1
Solution Type Neat
SMILES CC(=O)O.NC(=N)c1cn([C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O)c3N=C(N)NC(=O)c13
IUPAC Name acetic acid;2-amino-7-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-4-oxo-3H-pyrrolo[2,3-d]pyrimidine-5-carboximidamide
Industry Type Pharmaceutical
Molecular Weight (g/mol) 384.345
Quantity 1 mg
Formula Weight 384.139
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.