missing translation for 'onlineSavingsMsg'
Learn More

Arainosine, TRC

Product Code. 30134211
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134211 100 mg 100mg
30007167 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134211 Supplier Toronto Research Chemicals Supplier No. TRCA765200100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7013-16-3
Molecular Formula C10H12N4O5
Molecular Weight (g/mol) 268.23
Synonym 9-beta-D-Arabinofuranosyl-1,9-dihydro-6H-purin-6-one, 9-beta-D-Arabinofuranosylhypoxanthine, Ara-H, Arabinosylhypoxanthine, Hypoxanthine Arabinoside, NSC 405122
IUPAC Name 9-[(2R,3S,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1H-purin-6-one
SMILES OC[C@H]1O[C@H]([C@@H](O)[C@@H]1O)n2cnc3C(=O)NC=Nc23

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C10H12N4O5/c15-1-4-6(16)7(17)10(19-4)14-3-13-5-8(14)11-2-12-9(5)18/h2-4,6-7,10,15-17H,1H2,(H,11,12,18)/t4-,6-,7+,10-/m1/s1
Formula Weight 268.0808
Chemical Name or Material Arainosine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.