Learn More
Hirschmann™ AR-Glass Graduated Pipet
Click to view available options
Volume (Metric):
2 mL
5 mL
10 mL
25 mL
Graduations:
0.05 mL
0.1 mL
0.2 mL
Unit Size:
Pack of 12
Pack of 6
Description
- Main point ring graduation
- Cotton plug end
- Class AS, Conformity certified, with dated batch
Specifications
Specifications
| Description | Short form for tissue culture |
| Diameter (Metric) Outer | 8 mm |
| Material | Soda Lime Glass |
| For Use With (Application) | For tissue culture and working in cleanroom benches |
| Graduated | Yes |
| Graduation Color | Amber |
| Graduations | 0.2 mL |
| Graduation Type | Durable amber stain diffusion ink |
| Quantity | 12 Pack |
| Volume (Metric) | 25 mL |
| Show More |