Learn More
BRAND™ AR-Glas™ Stirring Rods
Glass stirring rods
Brand:  BRAND™ 135805
27.33 EUR valid until 2024-12-31
Use promo code "23264" to get your promotional price.
Additional Details : Weight : 0.42000kg
 View more versions of this product
Product Code. 10672242
  • 30.90 EUR / Pack of 50
Estimated Shipment: 17-06-2024
Specifications
5 mm
Fused closely at both ends
200 mm