missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aquaporin-5 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33420-20ul
This item is not returnable.
View return policy
Description
Aquaporin-5 Monoclonal antibody specifically detects Aquaporin-5 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| Aquaporin-5 | |
| Monoclonal | |
| Western Blot 1:1000 - 1:5000, ELISA Recommended starting concentration is 1 μg/mL | |
| AQP-5, aquaporin 5, aquaporin-5 | |
| A synthetic peptide corresponding to a sequence within amino acids 165-265 of human Aquaporin-5 (NP_001642.1).,, Sequence:, IGLSVTLGHLVGIYFTGCSMNPARSFGPAVVMNRFSPAHWVFWVGPIVGAVLAAILYFYLLFPNSLSLSERVAIIKGTYEPDEDWEEQREERKKTMELTTR | |
| 20 μL | |
| Cancer, Signal Transduction | |
| 362 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction