Learn More
Applied Biosystems™ MicroAmp™ Optical 384-Well Reaction Plate with Barcode
Product Code. 10005724 Shop All Applied Biosystems Products
Change view
Click to view available options
Includes:
1000 plates
50 plates
500 plates
Unit Size:
Each
Pack of 50
Pack of 500
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes unitSize
10005724 50 plates Pack of 50
10209594 500 plates Pack of 500
10505035 1000 plates Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price
Product Code. 10005724 Supplier Applied Biosystems™ Supplier No. 4309849
Only null left
Add to basket
Add to basket
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price
Designed to provide unmatched temperature accuracy and uniformity for fast, efficient PCR amplification
  • Constructed from a single rigid piece of polypropylene in a 384-well format
  • Designed to work with Applied Biosystems 384-well Real-Time PCR systems and thermal cyclers
  • Frosted to minimize interfering fluorescence from wells of cycling block
  • Screened to eliminate auto-fluorescent plates
  • Unique design provides barrier to ambient air to help ensure well-to-well temperature uniformity
  • Each reaction plate includes unique serialized, eight character number label (barcode) that is user-readable and machine-readable to prevent tracking errors
  • Platform: 3130 Genetic Analyzer, 3130xl Genetic Analyzer, 3500 Dx Genetic Analyzer, 3500 Genetic Analyzer, 3500xL Dx Genetic Analyzer, 3500xL Genetic Analyzer, 3730 DNA Analyzer, 3730xl DNA Analyzer, 7900HT System, GeneAmp™ 9700, QuantSt
PCR, PCR & Real-Time PCR, Real Time PCR (qPCR)
TRUSTED_SUSTAINABILITY
Specifications
Barcode Includes Barcode
Certifications/Compliance Certified DNA/RNase Free
Content And Storage Contains 50 plates.
May be stored at room temperature. Under proper storage conditions, plates may be stored indefinitely.
Packaging 50 Plates
Product Line MicroAmp
Color Optical
Product Type Microplate
For Use With (Equipment) Thermal Cycler, Genetic Analyzer, DNA Analyzer
Skirt Style Skirted
Material Polypropylene
Volume (Metric) Well 40 μL
No. of Wells 384
Includes 50 plates
Quantity 50 plates
Reaction Speed Fast or Standard
Shipping Condition Room Temperature