Description
- Durable design allows for continuous use with high-quality results
- Convenient, cost-saving alternative to MicroAmp Caps
- Patented one-piece design for convenience
- Reusable up to five times
- Manufactured from PCR-compatible rubber
- Non-sterile and autoclavable
- Platform: GeneAmp™ 9700
See user's manual or package insert for limited label license, and trademark information.
PCR, PCR & Real-Time PCR
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Certifications/Compliance | Certified DNA/RNase Free |
| Content And Storage | MicroAmp™ 96-Well Full Plate Cover is manufactured from PCR-compatible rubber. It is non-sterile, autoclavable and guaranteed reusable up to five times. |
| Packaging | 5 Covers |
| Product Line | MicroAmp |
| Color | Natural |
| Product Type | Microplate Lid |
| For Use With (Equipment) | 2720 Thermal Cycler, 6100 Thermal Cycler, GeneAmp 9700, 2720 Thermal Cycler |
| No. of Wells | 96 |
| Sterility | Non-sterile |
| For Use With (Application) | PCR |
| Show More |