missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This gene is a member of the apolipoprotein L gene family. The encoded protein is found in the cytoplasm, where it may affect the movement of lipids or allow the binding of lipids to organelles. Two transcript variants encoding the same protein have been found for this gene.
Specifications
Specifications
| Antigen | APOL2 |
| Applications | Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.53 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gene | APOL2 |
| Gene Accession No. | Q9BQE5 |
| Gene Alias | APOL II, APOL2, APOL3, Apolipoprotein L II, apolipoprotein L, 2, Apolipoprotein L2 |
| Gene Symbols | APOL2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?