Learn More
APOL1 Monoclonal Antibody (OTI1D8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812042S
This item is not returnable.
View return policy
Description
APOL1 Monoclonal antibody specifically detects APOL1 in Human samples. It is validated for Western BlotSpecifications
| APOL1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O14791 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 8542 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI1D8 | |
| Unconjugated | |
| APOL1 | |
| APOL; APO-L; APOL- I; APOL1; APOL-I; Apolipoprotein L; apolipoprotein L 1; apolipoprotein L, 1; Apolipoprotein L1; Apolipoprotein L-I; FSGS4; OTTHUMP00000028705 | |
| Human recombinant protein fragment corresponding to amino acids 28-339 of human APOL1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |