Learn More
APOL1 Monoclonal Antibody (OTI1D8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA812042S
Encompass_Preferred
 View more versions of this product
Product Code. 17556364
  • 242.00 EUR / 30µL
Description
APOL1 Monoclonal antibody specifically detects APOL1 in Human samples. It is validated for Western Blot
Specifications
Specifications
APOL1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
O14791
Mouse
Affinity chromatography
RUO
8542
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI1D8
Unconjugated
APOL1
APOL; APO-L; APOL- I; APOL1; APOL-I; Apolipoprotein L; apolipoprotein L 1; apolipoprotein L, 1; Apolipoprotein L1; Apolipoprotein L-I; FSGS4; OTTHUMP00000028705
Human recombinant protein fragment corresponding to amino acids 28-339 of human APOL1 produced in E.coli.
30 μL
Primary
Human
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions