missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APOBEC4 Monoclonal Antibody (OTI1F7), TrueMAB™, OriGene
Description
APOBEC4 Monoclonal antibody specifically detects APOBEC4 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | APOBEC4 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI1F7 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| Gene | PRELID2 |
| Gene Accession No. | Q8N945 |
| Gene Alias | 4933431M11Rik; APOBEC4; apolipoprotein B mRNA editing enzyme catalytic polypeptide like 4; apolipoprotein B mRNA editing enzyme catalytic polypeptide like 4 (putative); apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 4 (putative); apolipoprotein B mRNA-editing enzyme catalytic polypeptide-like 4; C1orf169; putative C->U-editing enzyme APOBEC-4 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?