missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ APOB48R Polyclonal Antibody
GREENER_CHOICE

Product Code. 16634095
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16634095 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16634095 Supplier Invitrogen™ Supplier No. PA599847

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total APOB48R.

Macrophage receptor that binds to the apolipoprotein B48 (APOB) of dietary triglyceride (TG)-rich lipoproteins (TRL) or to a like domain of APOB in hypertriglyceridemic very low density lipoprotein (HTG-VLDL). Binds and internalizes TRL when out of the context of the macrophage. May provide essential lipids to reticuloendothelial cells. Could also be involved in foam cell formation with elevated TRL and remnant lipoprotein (RLP). Mediates the rapid high-affinity uptake of chylomicrons (CM), HTG- VLDL, and trypsinized (tryp) VLDL devoid of APOE in vitro in macrophages.
TRUSTED_SUSTAINABILITY

Specifications

Antigen APOB48R
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene APOBR
Gene Accession No. Q0VD83
Gene Alias APOB100R; Apob48r; apoB-48R; Apobr; Apolipoprotein B receptor; apolipoprotein B100 receptor; apolipoprotein B-100 receptor; apolipoprotein B48 receptor; Apolipoprotein B-48 receptor; RGD1565169
Gene Symbols APOBR
Host Species Rabbit
Immunogen A synthesized peptide derived from human APOBR(Accession Q0VD83), corresponding to amino acid residues P1033-G1083.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55911
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.