missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apoatropine-N-oxide HCl, MP Biomedicals
Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 g
Unit Size:
50mg
Specifications
Specifications
| CAS | 4574-60-1 |
| Molecular Formula | C17H24ClNO4 |
| MDL Number | MFCD00035160 |
| Quantity | 1 g |
| Synonym | atropine-n-oxide hydrochloride, atropine n-oxide hydrochloride, atropine oxide hydrochloride usan, atropine oxide hydrochloride, prestwick_83, atropine oxide hcl, genatropine hydrochloride, aminoxytropine tropate hcl, dsstox_cid_25384 |
| InChI Key | QTRBWTMEQQSFLO-ICPLZUBISA-N |
| SMILES | [H+].[Cl-].C[N+]1([O-])[C@H]2CC[C@@H]1CC(C2)OC(=O)C(CO)C1=CC=CC=C1 |
| IUPAC Name | hydrogen (1R,5S)-3-[(3-hydroxy-2-phenylpropanoyl)oxy]-8-methyl-8-azabicyclo[3.2.1]octan-8-ium-8-olate chloride |
| Molecular Weight (g/mol) | 341.83 |
| PubChem CID | 6419888 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction