missing translation for 'onlineSavingsMsg'
Learn More

Antioxidant 1098, TRC

Product Code. 30133063
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133063 10 g 10g
30138030 5 g 5g
17937804 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133063 Supplier Toronto Research Chemicals Supplier No. TRCA69752310G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23128-74-7
Molecular Formula C40 H64 N2 O4
Molecular Weight (g/mol) 636.95
Synonym Hydrocinnamamide, N,N'-hexamethylenebis[3,5-di-tert-butyl-4-hydroxy- (8CI), N,N'-1,6-Hexanediylbis[3,5-bis(1,1-dimethylethyl)-4-hydroxybenzenepropanamide, 1,6-Bis(3,5-di-tert-butyl-4-hydroxyhydrocinnamido)hexane, 1,6-Bis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionamido]hexane, 1,6-Bis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionylamino]hexane, 1,6-Bis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propylamido]hexane, 3,3'-Bis(3,5-di-tert-butyl-4-hydroxyphenyl)-N,N'-hexamethylenedipropionamide, Antioxidant 1096, Antioxidant 1098, Chemnox 1098, GX 2921, HD 98, Heat stabilizer 1096, IX 1098, Irg 1098, Irganox 1090, Irganox 1098, Irganox 98, Irganox B 1096, KY 1098, Lowinox HD 98, N,N'-1,6-Hexamethylenebis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionamide, N,N'-Bis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionyl]hexamethylenediamine, N,N'-Bis[3-(3,5-tert-butyl-4-hydroxyphenyl)propionyl]hexamethylenediamine, N,N'-Bis[3-(3',5'-di-tert-butyl-4'-hydroxyphenyl)propionyl]hexamethylenediamine, N,N'-Hexamethylenebis(3,5-di-tert-butyl-4-hydroxybenzenepropionamide), N,N'-Hexamethylenebis(3,5-di-tert-butyl-4-hydroxyhydrocinnamamide), N,N'-Hexamethylenebis(3,5-di-tert-butyl-4-hydroxyhydrocinnamide), N,N'-Hexamethylenebis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionamide], N,N'-Hexane-1,6-diylbis[3,5-ditert-butyl-4-hydroxyphenylpropionamide)], N,N'-Hexane-1,6-diylbis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionamide], TTAD
IUPAC Name 3-(3,5-ditert-butyl-4-hydroxyphenyl)-N-[6-[3-(3,5-ditert-butyl-4-hydroxyphenyl)propanoylamino]hexyl]propanamide
SMILES CC(C)(C)c1cc(CCC(=O)NCCCCCCNC(=O)CCc2cc(c(O)c(c2)C(C)(C)C)C(C)(C)C)cc(c1O)C(C)(C)C

Specifications

Quantity 10 g
InChI Formula InChI=1S/C40H64N2O4/c1-37(2,3)29-23-27(24-30(35(29)45)38(4,5)6)17-19-33(43)41-21-15-13-14-16-22-42-34(44)20-18-28-25-31(39(7,8)9)36(46)32(26-28)40(10,11)12/h23-26,45-46H,13-22H2,1-12H3,(H,41,43)(H,42,44)
Formula Weight 636.4866
Chemical Name or Material Antioxidant 1098
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.